Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBFB Antibody (PCRP-CBFB-1F6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314058IR
This item is not returnable.
View return policy
Description
CBFB Monoclonal antibody specifically detects CBFB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| CBFB | |
| Monoclonal | |
| DyLight 755 | |
| CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta | |
| Recombinant full-length human CBFB protein (Uniprot: Q13951) | |
| 0.1 mL | |
| DNA Replication Transcription Translation and Splicing | |
| 865 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-CBFB-1F6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction