Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBL Antibody (3B12), Novus Biologicals™
SDP

Catalog No. NBP237574 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237574 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237574 Supplier Novus Biologicals Supplier No. NBP237574
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CBL Monoclonal specifically detects CBL in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CBL
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3B12
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin
Formulation Ascites with 0.03% Sodium Azide
Gene Alias Cas-Br-M (murine) ecotropic retroviral transforming sequence, Casitas B-lineage lymphoma proto-oncogene, CBL2E3 ubiquitin-protein ligase CBL, EC 6.3.2, EC 6.3.2.-, oncogene CBL2, Proto-oncogene c-Cbl, RING finger protein 55, RNF55C-CBL, Signal transduction protein CBL
Gene Symbols CBL
Host Species Mouse
Immunogen Purified recombinant fragment of human C-CBL expressed in E. Coli.
Molecular Weight of Antigen 120 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 867
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.