Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cbl-b Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554107 Supplier Invitrogen™ Supplier No. PA554107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNRLSRHIHH VESVPSRDPP MPLEAWCPRD VFGTNQLVGC RLLGEGSPKP GITASSNVNG RHSRVGSDPV LMRKHRRHDL PLEGAKVFSN GHLGSEEYDV PPRLSPPPPV TTLLPSIKCT GPLA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cbl-b
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CBLB
Gene Accession No. Q13191
Gene Alias AI429560; AI851073; Cas-Br-M (murine) ecotropic retroviral transforming sequence b; Cas-Br-M (murine) ectropic retroviral transforming sequence b; Casitas B-lineage lymphoma b; casitas B-lineage lymphoma proto-oncogene b; Cbl proto-oncogene B; Cbl proto-oncogene B, E3 ubiquitin protein ligase; Cbl proto-oncogene, E3 ubiquitin protein ligase B; CBLB; Cbl-b; E3 ubiquitin-protein ligase CBL-B; Nbla00127; RING finger protein 56; RING-type E3 ubiquitin transferase CBL-B; RNF56; SH3-binding protein CBL-B; signal transduction protein CBL-B
Gene Symbols CBLB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cbl-b. Recombinant protein control fragment (Product #RP-91840).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 868
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.