Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cbl-c Antibody (OTI3B4), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271755PECY55
Description
Cbl-c Monoclonal specifically detects Cbl-c in Human samples. It is validated for Flow Cytometry.Specifications
Cbl-c | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CBLC | |
Recombinant protein fragment corresponding to amino acids 244-474 of human Cbl-c (NP_036248) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Flow Cytometry | |
OTI3B4 | |
Flow Cytometry | |
Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
23624.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction