Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cbl-c Antibody (OTI4E3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271756
Description
Cbl-c Monoclonal specifically detects Cbl-c in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Cbl-c | |
Monoclonal | |
LYOPH | |
Western Blot 1:500, Flow Cytometry 1:100, CyTOF-ready | |
Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C | |
Mouse | |
52.3 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, CyTOF | |
OTI4E3 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
CBLC | |
Human recombinant protein fragment corresponding to amino acids 244-474 of human CBLC(NP_036248) produced in HEK293T cell. | |
Affinity Purified | |
Stem Cell Markers | |
23624 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction