Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBL Rabbit anti-Human, Mouse, Rat, APC, Clone: E1, Abnova™

Catalog No. 89335672 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-672 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-672 Supplier Abnova Corporation Supplier No. MAB18838
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CBL Monoclonal antibody specifically detects CBL in Human, Mouse, Rat samples. It is validated for Flow Cyt.

Specifications

Antigen CBL
Applications Flow Cytometry
Classification Monoclonal
Clone E1
Conjugate APC
Description Cas-Br-M (murine) ecotropic retroviral transforming sequence
Gene CBL
Gene Alias C-CBL, CBL2, RNF55
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y700 of human CBL
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 867
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.