Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBLB Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149920JF549
Description
CBLB Polyclonal antibody specifically detects CBLB in Human samples. It is validated for Western BlotSpecifications
CBLB | |
Polyclonal | |
Western Blot | |
Cas-Br-M (murine) ecotropic retroviral transforming sequence b, Cas-Br-M (murine) ectropic retroviral transforming sequence b, Casitas B-lineage lymphoma proto-oncogene b, DKFZp686J10223, DKFZp779A0729, DKFZp779F1443, E3 ubiquitin-protein ligase CBL-B, EC 6.3.2, EC 6.3.2.-, FLJ41152, RING finger protein 56, RNF56FLJ36865, SH3-binding protein CBL-B, Signal transduction protein CBL-B | |
The immunogen for this product maps to a region between residue 932 and 982 of human Cas-Br-M (murine) Ecotropic Retroviral Transforming Sequence b using the numbering given in entry NP_733762.2 (GeneID 868). | |
0.1 mL | |
Signal Transduction | |
868 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction