Learn More
CBLC Mouse anti-Human, Clone: OTI3B4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505071

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Acts as an E3 ubiquitin-protein ligase, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes, and then transfers it to substrates promoting their degradation by the proteasome. Functionally coupled with the E2 ubiquitin-protein ligases UB2D1, UB2D2 and UB2D3. Regulator of EGFR mediated signal transduction; upon EGF activation, ubiquitinates EGFR. Isoform 1, but not isoform 2, inhibits EGF stimulated MAPK1 activation. Promotes ubiquitination of SRC phosphorylated at ′Tyr-419′. In collaboration with CD2AP may act as regulatory checkpoint for Ret signaling by modulating the rate of RET degradation after ligand activation; CD2AP converts it from an inhibitor to a promoter of RET degradation; the function limits the potency of GDNF on neuronal survival.Specifications
CBLC | |
Monoclonal | |
Unconjugated | |
CBLC | |
CBL-3, CBL-SL, RNF57 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23624 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI3B4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9ULV8 | |
CBLC | |
Human recombit protein fragment corresponding to amino acids 244-474 of human CBLC produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.