Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBLL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554503 Supplier Invitrogen™ Supplier No. PA554503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKHSNLITVP IQDDSNSGAR EPPPPAPAPA HHHPEYQGQP VVSHPHHIMP PQQHYAPPPP PPPPISHPMP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes an E3 ubiquitin-ligase for the E-cadherin complex and mediates its ubiquitination, endocytosis, and degradation in the lysosomes. The encoded protein contains a RING-finger domain and is also thought to have a role in control of cell proliferation. A related pseudogene has been identified on chromosome X. Alternative splicing results in a non-coding transcript variant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBLL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CBLL1
Gene Accession No. Q75N03
Gene Alias AI467391; Cas-Br-M (murine) ecotropic retroviral transforming sequence-like 1; Casitas B-lineage lymphoma-like 1; casitas B-lineage lymphoma-transforming sequence-like protein 1; Cbl proto-oncogene like 1; Cbl proto-oncogene like 1, E3 ubiquitin protein ligase; Cbl proto-oncogene, E3 ubiquitin protein ligase-like 1; Cbl proto-oncogene-like 1, E3 ubiquitin protein ligase; CBLL1; c-Cbl-like; c-Cbl-like protein 1; E3 ubiquitin-protein ligase Hakai; E-cadherin binding protein E7; Hakai; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase Hakai; RING finger protein 188; RING-type E3 ubiquitin transferase Hakai; RNF188
Gene Symbols CBLL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CBLL1. Recombinant protein control fragment (Product #RP-92641).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79872
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.