Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBP/KAT3A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298753AF700
Description
CBP/KAT3A Polyclonal specifically detects CBP/KAT3A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CBP/KAT3A | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
CBPRTS, CREB binding protein, CREB-binding protein, EC 2.3.1.48, KAT3A, RSTS, Rubinstein-Taybi syndrome | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human CBP/KAT3A. | |
0.1 mL | |
Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
1387 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction