Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBP/KAT3A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100382PCP
Description
CBP/KAT3A Polyclonal antibody specifically detects CBP/KAT3A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
CBP/KAT3A | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
CBPRTS, CREB binding protein, CREB-binding protein, EC 2.3.1.48, KAT3A, RSTS, Rubinstein-Taybi syndrome | |
Maps to a region between residues 2375 and 2425 of human CREB Binding Protein using the numbering given in entry NP_004371.1 (GeneID 1387). | |
0.1 mL | |
Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
1387 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction