Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBP/KAT3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100381
Description
CBP/KAT3A Polyclonal specifically detects CBP/KAT3A in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.Specifications
CBP/KAT3A | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
CBPRTS, CREB binding protein, CREB-binding protein, EC 2.3.1.48, KAT3A, RSTS, Rubinstein-Taybi syndrome | |
Rabbit | |
265 kDa | |
100 μL | |
Stem Cell Signaling Pathway | |
1387 | |
Human | |
IgG |
Western Blot, Immunoprecipitation, ChIP Assay | |
1.0 mg/mL | |
Western Blot 1:1000 - 1:25000, Immunoprecipitation 1-4 ug/mg of lysate, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Q92793 | |
CREBBP | |
The immunogen recognized by this antibody maps to a region between residues 975 and 1025 of human CREB Binding Protein using the numbering given in entry NP_004371.1 (GeneID 1387). | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction