Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBP Monoclonal Antibody (NM11), Biotin, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA513631

Catalog No. PIMA513631


Only null left
Add to Cart

Description

Description

MA5-13631 targets p300/CBP (CREB-Binding Protein) in IF and IHC (P) applications and shows reactivity with Human, Mink, mouse, Non-human primate, and Rat samples. The MA5-13631 immunogen is e1A affinity purified p300 protein.

CBP for CREB-binding protein is a 265 kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CBP
Monoclonal
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P45481, Q6JHU9, Q92793
CREBBP
E1A affinity purified p300 protein
500 μL
Primary
Human, Mink, Mouse, Non-human Primate, Rat
Antibody
IgG2b κ
Immunohistochemistry (Paraffin)
NM11
Biotin
CREBBP
AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS
Mouse
Protein A
RUO
12914, 1387, 54244
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.