Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR1 Mouse anti-Human, Clone:2C9-B12-C4, Abnova™

Catalog No. 89333880 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-880 Supplier Abnova Corporation Supplier No. MAB17658

Mouse monoclonal antibody raised against recombinant human CBR1.

Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q. [provided by RefSeq]

Specifications

Antigen CBR1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2C9-B12-C4
Conjugate Unconjugated
Description carbonyl reductase 1
Dilution Immunocytochemistry (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.2% sodium azide)
Gene CBR1
Gene Alias CBR, SDR21C1, hCBR1
Host Species Mouse
Immunogen Recombinant protein corresponding to a fragment of human CBR1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 873
Test Specificity This antibody detects endogenous levels of CBR1 and does not cross-react with related proteins.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.