Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106929 Supplier Invitrogen™ Supplier No. PA5106929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CBR1.

Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CBR1
Gene Accession No. P16152, P47727, P48758
Gene Alias 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NADP(+)]; 20-beta-hydroxysteroid dehydrogenase; AW261796; carbonyl reductase (NADPH) 1; carbonyl reductase [NADPH] 1; Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1); carbonyl reductase 1; CBR; Cbr 1; Cbr1; CR; CRN; hCBR1; NADPH-dependent carbonyl reductase 1; prostaglandin 9-ketoreductase; Prostaglandin-E(2) 9-reductase; SDR21C1; Short chain dehydrogenase/reductase family 21C member 1; short chain dehydrogenase/reductase family 21C, member 1
Gene Symbols CBR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CBR1(Accession P16152), corresponding to amino acid residues E246-W277.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12408, 29224, 873
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.