Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBR1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $536.00
Specifications
Antigen | CBR1 |
---|---|
Applications | Western Blot, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
CBR1 Polyclonal specifically detects CBR1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
CBR1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
15-hydroxyprostaglandin dehydrogenase [NADP+], carbonyl reductase (NADPH) 1, carbonyl reductase (NADPH) 1, EC 1.1.1.18410EC 1.1.1.197,15-hydroxyprostaglandin dehydrogenase, carbonyl reductase [NADPH] 1, carbonyl reductase 1, CBR, CRN, EC 1.1.1.184, EC 1.1.1.189, hCBR1, NADPH-dependent carbonyl reductase 1, Prostaglandin 9-ketoreductase, Prostaglandin-E(2) 9-reductase, SDR21C1, short chain dehydrogenase/reductase family 21C, member 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human CBR1 (NP_001748.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
873 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title