Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBR1 Rabbit anti-Human, Mouse, Rat, Clone: 3A9O5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CBR1 |
---|---|
Clone | 3A9O5 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
CBR1 Monoclonal antibody specifically detects CBR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CBR1 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
15-hydroxyprostaglandin dehydrogenase [NADP+], carbonyl reductase (NADPH) 1, carbonyl reductase (NADPH) 1, EC 1.1.1.18410EC 1.1.1.197,15-hydroxyprostaglandin dehydrogenase, carbonyl reductase [NADPH] 1, carbonyl reductase 1, CBR, CRN, EC 1.1.1.184, EC 1.1.1.189, hCBR1, NADPH-dependent carbonyl reductase 1, Prostaglandin 9-ketoreductase, Prostaglandin-E(2) 9-reductase, SDR21C1, short chain dehydrogenase/reductase family 21C, member 1 | |
A synthetic peptide corresponding to a sequence within amino acids 178-277 of human CBR1 (P16152). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3A9O5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
873 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title