Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBR3 Antibody (OTI1G6), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270343PCP
Description
CBR3 Monoclonal antibody specifically detects CBR3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
CBR3 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate | |
Purified |
Western Blot, Immunohistochemistry | |
OTI1G6 | |
Western Blot, Immunohistochemistry | |
carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2 | |
Full length human recombinant protein of human CBR3(NP_001227) produced in HEK293T cell. | |
0.1 mL | |
Lipid and Metabolism | |
874 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction