Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBR3 Monoclonal Antibody (OTI1G6)
GREENER_CHOICE

Catalog No. PIMA525626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525626 Supplier Invitrogen™ Supplier No. MA525626
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CBR3 Monoclonal Antibody for Western Blot, IHC (P)

Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBR3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G6
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CBR3
Gene Accession No. O75828, Q8K354
Gene Alias 1110001J05Rik; C81353; carbonyl reductase (NADPH) 3; carbonyl reductase [NADPH] 3; carbonyl reductase 3; Cbr3; epididymis secretory protein Li 25; hCBR3; HEL-S-25; NADPH-dependent carbonyl reductase 3; SDR21C2; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2
Gene Symbols CBR3
Host Species Mouse
Immunogen Full length human recombinant protein of CBR3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109857, 304078, 487748, 874
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.