Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBR3 Mouse anti-Human, Mouse, Clone: 7.00E+08, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | CBR3 |
---|---|
Clone | 700000000 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1013, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25940150
![]() |
Novus Biologicals
NBP25940150UL |
50 μL |
Each for $340.50
|
|
|||||
NBP259401X
![]() |
Novus Biologicals
NBP259401100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
CBR3 Monoclonal specifically detects CBR3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CBR3 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Lipid and Metabolism | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
874 | |
Recombinant human CBR3 (1-277aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
700000000 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1013, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2 | |
CBR3 | |
IgG1 κ | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title