Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553800 Supplier Invitrogen™ Supplier No. PA553800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDPMPFDIKA EMTLKTNFFA TRNMCNELLP IMKPHGRVVN ISSLQCLRAF ENCSEDLQER FHSETLTEGD LVDLMK Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.

Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBR3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CBR3
Gene Accession No. O75828, Q8K354
Gene Alias 1110001J05Rik; C81353; carbonyl reductase (NADPH) 3; carbonyl reductase [NADPH] 3; carbonyl reductase 3; Cbr3; epididymis secretory protein Li 25; hCBR3; HEL-S-25; NADPH-dependent carbonyl reductase 3; SDR21C2; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2
Gene Symbols CBR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CBR3. Recombinant protein control fragment (Product #RP-91772).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109857, 304078, 874
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.