Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121115 Supplier Invitrogen™ Supplier No. PA5121115
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SW480, NCI-H460, MCF7, BT-474, Mouse lung, Mouse brain, Mouse ovary, Rat brain. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: TRNMCNELLP IMKPHGRVVN ISSLQCLRAF ENCSEDLQER FHSETLTEGD LVDLMKKFVE DTKNEVHERE GWPNSPYGVS KLGVTVLSRI LARRLDEKRK ADRILVNACC PGPVKTDMDG KDSIRTVEEG AETPVYLALL PPDATEPQGQ LVHDKVVQNW.

Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBR3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CBR3
Gene Accession No. O75828, Q8K354
Gene Alias 1110001J05Rik; C81353; carbonyl reductase (NADPH) 3; carbonyl reductase [NADPH] 3; carbonyl reductase 3; Cbr3; epididymis secretory protein Li 25; hCBR3; HEL-S-25; NADPH-dependent carbonyl reductase 3; SDR21C2; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2
Gene Symbols CBR3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 118-277 of human CBR3 (NP_001227.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109857, 304078, 874
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.