Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen CBR4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179860
SDP
View Documents
Novus Biologicals
NBP179860
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

CBR4 Polyclonal specifically detects CBR4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CBR4
Polyclonal
Rabbit
Lipid and Metabolism
3-oxoacyl-[acyl-carrier-protein] reductase, carbonic reductase 4, carbonyl reductase 4, carbonyl reductase family member 4, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.100, FLJ14431, Quinone reductase CBR4, SDR45C1, short chain dehydrogenase/reductase family 45C, member 1
CBR4
IgG
25 kDa
Western Blot
Unconjugated
RUO
NP_116172
84869
Synthetic peptide directed towards the N terminal of human CBR4The immunogen for this antibody is CBR4. Peptide sequence RKGYRLAVIARNLEGAKAAAGDLGGDHLAFSCDVAKEHDVQNTFEELEKH.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.