Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR4 Monoclonal Antibody (OTI3C10), TrueMAB™, OriGene
SDP

Catalog No. 502543978
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3978 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3978 Supplier OriGene Supplier No. CF810992
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CBR4 Monoclonal antibody specifically detects CBR4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CBR4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3C10
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ASH2L
Gene Accession No. Q91X20, Q9UBL3
Gene Alias 3-ketoacyl-[acyl-carrier-protein] reductase beta subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; A730083J17Rik; BC009118; carbonic reductase 4; carbonyl reductase 4; carbonyl reductase family member 4; Cbr4; KAR beta subunit; Quinone reductase CBR4; SDR45C1; Short chain dehydrogenase/reductase family 45C member 1; short chain dehydrogenase/reductase family 45C, member 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 347-628 of human ASH2L produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23808, 290829, 9070
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.