Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557977 Supplier Invitrogen™ Supplier No. PA557977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLVNAAGINR DGLLVRTKTE DMVSQLHTNL LGSMLTCKAA MRTMIQQQGG SIVNVGSIVG LKGNSGQSVY SAS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.

The heterotetramer with HSD17B8 has NADH-dependent 3-ketoacyl-acyl carrier protein reductase activity, and thereby plays a role in mitochondrial fatty acid biosynthesis. Within the heterotetramer, HSD17B8 binds NADH; CBR4 binds NADPD. The homotetramer has NADPH-dependent quinone reductase activity. Both homotetramer and the heterotetramer have broad substrate specificity and can reduce 9,10-phenanthrenequinone, 1,4-benzoquinone and various other o-quinones and p-quinones (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBR4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CBR4
Gene Accession No. Q8N4T8
Gene Alias 3-ketoacyl-[acyl-carrier-protein] reductase beta subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; A730083J17Rik; BC009118; carbonic reductase 4; carbonyl reductase 4; carbonyl reductase family member 4; Cbr4; KAR beta subunit; Quinone reductase CBR4; SDR45C1; Short chain dehydrogenase/reductase family 45C member 1; short chain dehydrogenase/reductase family 45C, member 1
Gene Symbols CBR4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CBR4. Recombinant protein control fragment (Product #RP-96684).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.