Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728962 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8962 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8962 Supplier Proteintech Group Inc Supplier No. 137251AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The heteroteramer with HSD17B8 has NADH-dependent 3-ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria. The homotetramer may act as NADPH-dependent quinone reductase. Has broad substrate specificity and reduces 9,10-phethrenequinone, 1,4-benzoquinone and various other o-quinones and p-quinones (in vitro).

Specifications

Antigen CBR4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CBR4
Gene Accession No. Q7TS56, Q8N4T8, Q91VT4
Gene Alias carbonyl reductase 4, CBR4, Quinone reductase CBR4, SDR45C1
Gene Symbols Cbr4
Host Species Rabbit
Immunogen CBR4 Fusion Protein Ag4678
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234309, 359725, 84869
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.