Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CBS HEK293 Cell Transient Overexpression Lysate (Nondenatured)

Catalog No. 89938188 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-188 Supplier Abnova™ Supplier No. L043T6
Add to Cart
Add to Cart

Human overexpression lysate

Specifications

Cell Line HEK293
For Use With (Application) Immunoprecipitation, Western Blot
Description CBS HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Gene Accession No. NM_000071 ;NM_000071
Lysis Buffer Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
Molecular Weight (g/mol) Theoretical M.W. 61kDa
Plasmid pCMV-CBS full length ;pCMV-CBS full length
Quality Control Testing Transient overexpression cell lysate was tested with anti-CBS antibody (Mfr. No. H00000875-M01) by Western Blot.
Quantity 100 μg
Storage Buffer In modified RIPA Lysis Buffer. ;In modified RIPA Lysis Buffer.
Host Species Human
Concentration 2 mg/mL, 2 mg/mL
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing. ;Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.