Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBS Recombinant Rabbit Monoclonal Antibody (JE40-99)
GREENER_CHOICE

Catalog No. PIMA534826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534826 Supplier Invitrogen™ Supplier No. MA534826
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: 293, human liver tissue lysate, 293T.

Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBS
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE40-99
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CBS
Gene Accession No. P35520
Gene Alias AI047524; AI303044; Beta-thionase; Cbs; CBSL; cystathionine beta synthase; cystathionine beta-synthase; cystathionine beta-synthase; LOW QUALITY PROTEIN: cystathionine beta-synthase; Cystathionine beta-synthase-like protein; cystathionine-beta-synthase; hemoprotein H-450; HIP4; LOC100057612; methylcysteine synthase; serine sulfhydrase
Gene Symbols CBS
Host Species Rabbit
Immunogen Recombinant protein within Human CBS aa 360-551.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 875
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.