Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Scientific Dual Cool Mini-Vertical PAGE/Blotting System w/ Multi Gel Casting Chamber
SCI_ED

Catalog No. S05760 Shop All CBS Scientific Products
Change view
Click to view available options
Device Holds:
0.75 mm x 10-well Precast Gel
0.75 mm x 14-well Precast Gel
1 mm x 10-well Precast Gel
1 mm x 14-well Precast Gel
1.5 mm x 10-well Precast Gel
1.5 mm x 14-well Precast Gel
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Device Holds
S05760 0.75 mm x 10-well Precast Gel
S05761 0.75 mm x 14-well Precast Gel
S05762 1 mm x 10-well Precast Gel
S05763 1 mm x 14-well Precast Gel
S05765 1.5 mm x 14-well Precast Gel
S05764 1.5 mm x 10-well Precast Gel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S05760 Supplier CBS Scientific Supplier No. DCXMHC7510

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

DCX-700 with Multiple Gel Casting Chamber Bundle DCX-MHC.

  • Uniquely designed, multipurpose, easy-to-use Dual Cool System can accommodate both slab gel electrophoresis and Western blotting within the same unit without the purchase of additional modules
  • The Dual Cool System provides the capability of running 2 gels or 2 blots simultaneously under identical temperature controlled buffer conditions
  • DCX-700 Dual Cool Electrophoresis System includes lower reservoir, core, 2 blotting cassettes, 2 freezer blocks, safety cover with attached leads, single gel adapter plate
  • Each Bundle includes a Dual Cool DCX-700 and 4- place Multiple Gel Casting Chamber with lid, 4 sets notched glass plate sets, 4 sets spacers, 4 combs and polycarbonate barrier sheets

DCX-700

  • Molded system is designed primarily for use with a variety of precast gels ranging in size from 10cm x 10cm to 10cm x 9cm
  • Alternatively, hand-cast gels can be run with the use of accessory combs, plates, and spacers
  • Hinged core design allows easy loading and removal of gel or blotting cassettes
  • Freezer blocks fit into receptacles inside of unit for optional cooling
  • Electrophoretic transfers do not require the purchase of separate transfer modules
  • Color-coded anode and cathode safety features on all components
  • Clear tank allows for monitoring of sample progress
  • Platinum electrodes

SDS-PAGE, Electroblotting (Western), Acrylamide nucleic acid separations

Certifications

CE marked

Specifications

Ambient Temperature (English) 140°F
Ambient Temperature (Metric) 60°C
Certifications/Compliance EN61010-1-1993 (IEC1010-1), CE
Device Holds 0.75 mm x 10-well Precast Gel
Type Dual Cool Mini Vertical PAGE/Blotting System
Includes 4 Place multiple gel casting chamber bundle, 4 sets of multi caster notched Glass plates, 4 sets of multi caster 0.75mm spacers and four 0.75mm x 10-well combs
Volume (English) Buffer 13.52 to 27.38 oz.
Volume (Metric) Buffer 400 to 810 mL
Dimensions (L x W x H) 5.98 x 6.49 x 8.5 in. (15.2 x 16.5 x 21.6 cm)
Length (English) 5.98 in.
Length (English) Gel 3.93, 3.54, 3.14 in.
Length (Metric) 15.2 cm
Length (Metric) Gel 10, 9, 8 cm
Width (English) 6.49 in.
Width (English) Gel 3.93 in.
Width (Metric) 16.5 cm
Width (Metric) Gel 10 cm
Height (English) 8.5 in.
Height (Metric) 21.6 cm
Voltage 300 V
Wattage 30 w
Amperage 190 mA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.