Learn More
CBS Scientific Dual Cool Mini-Vertical PAGE/Blotting System w/ Multi Gel Casting Chamber
Shop All CBS Scientific Products

Description
- Uniquely designed, multipurpose, easy-to-use Dual Cool System can accommodate both slab gel electrophoresis and Western blotting within the same unit without the purchase of additional modules
- The Dual Cool System provides the capability of running 2 gels or 2 blots simultaneously under identical temperature controlled buffer conditions
- DCX-700 Dual Cool Electrophoresis System includes lower reservoir, core, 2 blotting cassettes, 2 freezer blocks, safety cover with attached leads, single gel adapter plate
- Each Bundle includes a Dual Cool DCX-700 and 4- place Multiple Gel Casting Chamber with lid, 4 sets notched glass plate sets, 4 sets spacers, 4 combs and polycarbonate barrier sheets
DCX-700
- Molded system is designed primarily for use with a variety of precast gels ranging in size from 10cm x 10cm to 10cm x 9cm
- Alternatively, hand-cast gels can be run with the use of accessory combs, plates, and spacers
- Hinged core design allows easy loading and removal of gel or blotting cassettes
- Freezer blocks fit into receptacles inside of unit for optional cooling
- Electrophoretic transfers do not require the purchase of separate transfer modules
- Color-coded anode and cathode safety features on all components
- Clear tank allows for monitoring of sample progress
- Platinum electrodes
SDS-PAGE, Electroblotting (Western), Acrylamide nucleic acid separations
Certifications
CE marked
Specifications
Specifications
| Ambient Temperature (English) | 140°F |
| Ambient Temperature (Metric) | 60°C |
| Certifications/Compliance | EN61010-1-1993 (IEC1010-1), CE |
| Device Holds | 0.75 mm x 14-well Precast Gel |
| Type | Dual Cool Mini Vertical PAGE/Blotting System |
| Includes | 4 Place multiple gel casting chamber bundle, 4 sets of multi caster notched Glass plates, 4 sets of multi caster 0.75mm spacers and four 0.75mm x 14-well combs |
| Volume (English) Buffer | 13.52 to 27.38 oz. |
| Volume (Metric) Buffer | 400 to 810 mL |
| Dimensions (L x W x H) | 5.98 x 6.49 x 8.5 in. (15.2 x 16.5 x 21.6 cm) |
| Length (English) | 5.98 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.