Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBWD1 Antibody (OTI4D4), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272027PE
Description
CBWD1 Monoclonal specifically detects CBWD1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
CBWD1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
CBWD1 | |
Full length recombinant protein of human CBWD1 (NP_060961) produced in HEK293T cells. | |
0.1 mL | |
55871.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI4D4 | |
Flow Cytometry | |
Cobalamin synthase W domain-containing protein 1, cobalamin synthetase W domain-containing protein 1, COBP, COBW domain containing 1, COBW domain-containing protein 1, COBW-like protein, NPC-A-6, NPC-A-6 COBW domain-containing protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat, Canine, Primate, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction