Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBWD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113110 Supplier Invitrogen™ Supplier No. PA5113110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CBWD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The CBWD2 gene, a member of the CobW domain-containing (CBWD) gene family, encodes a protein involved in vitamin B12 synthesis. This gene is part of a conserved pathway crucial for cobalamin biosynthesis, typically in microorganisms. In dinoflagellates, the CBWD gene family, including CBWD2, has been identified to play a role in the synthesis and regulation of vitamin B12, essential for various metabolic processes. The CobW domain is implicated in interactions with cobalt, a vital component of the vitamin B12 molecule, suggesting a regulatory or structural role in cobalamin assembly. Research on the CBWD2 gene and its protein products can provide insights into the complex mechanisms of vitamin B12 biosynthesis and its evolutionary significance in eukaryotic organisms, possibly opening avenues for biotechnological applications in vitamin B12 production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBWD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CBWD2
Gene Accession No. Q8IUF1
Gene Alias CBWD2; Cobalamin synthase W domain-containing protein 2; cobalamin synthetase W domain-containing protein 2; COBW domain containing 2; COBW domain-containing protein 2
Gene Symbols CBWD2
Host Species Rabbit
Immunogen Recombinant Human COBW domain-containing protein 2 protein (96-395AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150472
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.