Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBWD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549247
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549247 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549247 Supplier Invitrogen™ Supplier No. PA549247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CBWD6 Polyclonal Antibody for Western Blot

Zinc chaperone that directly transfers zinc cofactor to target metalloproteins, thereby activating them (By similarity). Catalyzes zinc insertion into the active site of methionine aminopeptidase METAP1, which function to cleave the initiator methionine from polypeptides during or after protein translation. Mechanistically, the N-terminal psi-PxLVp motif binds to the C6H2-type zinc finger of inactive form of METAP1 (By similarity). After formation of the docked complex, zinc is transferred from the CXCC motif in the GTPase domain of ZNG1F to the zinc binding site in the peptidase domain of METAP1 in a process requiring GTP hydrolysis (By similarity). GTP/GDP exchange is required for release of active METAP1 (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBWD6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CBWD6
Gene Accession No. Q4V339
Gene Alias CBWD6; CBWD7; Cobalamin synthase W domain-containing protein 6; cobalamin synthetase W domain-containing protein 6; COBW domain containing 6; COBW domain-containing protein 6
Gene Symbols CBWD6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 193-221 amino acids from the Central region of human CBWD6.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.