Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298756IR
Description
CBX1 Polyclonal specifically detects CBX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CBX1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox protein homolog 1, Heterochromatin protein 1 homolog beta, heterochromatin protein 1-beta, Heterochromatin protein p25, heterochromatin protein p25 beta, HP1 beta, HP1 beta homolog, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, p25beta | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CBX1. | |
0.1 mL | |
Chromatin Research | |
10951 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction