Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cbx2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP140342AF488
Description
Cbx2 Polyclonal antibody specifically detects Cbx2 in Human samples. It is validated for Western BlotSpecifications
Cbx2 | |
Polyclonal | |
Western Blot | |
CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog | |
The immunogen for this product maps to a region between residue 482 and 532 of human chromobox homolog 2 using the numbering given in entry NP_005180.1 (GeneID 84733). | |
0.1 mL | |
Chromatin Modifiers | |
84733 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction