Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cbx2 Antibody (SR2008), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32212550UL
Description
Cbx2 Monoclonal antibody specifically detects Cbx2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
Cbx2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR2008 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:150 | |
CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog | |
A synthesized peptide derived from human Cbx2 (Uniprot #: Q14781) | |
50 μg | |
Chromatin Modifiers | |
84733 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction