Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX3 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298757APC
Description
CBX3 Polyclonal antibody specifically detects CBX3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
CBX3 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox protein homolog 3, Drosophila), HECH, Heterochromatin protein 1 homolog gamma, heterochromatin protein HP1 gamma, heterochromatin-like protein 1, HP1 gamma, HP1 gamma homolog, HP1-GAMMA, Modifier 2 protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CBX3. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
11335 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction