Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX4 Antibody (6C5G3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | CBX4 |
---|---|
Clone | 6C5G3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
CBX4 Monoclonal specifically detects CBX4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
CBX4 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Apoptosis | |
PBS with 0.05% Sodium Azide | |
8535 | |
Purified recombinant fragment of human CBX4 (AA: 397-514) expressed in E. Coli. | |
Primary | |
61.4 kDa |
6C5G3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
chromobox homolog 4, chromobox homolog 4 (Drosophila Pc class), Chromobox protein homolog 4, Drosophila), E3 SUMO-protein ligase CBX4, hPC2, NS5ATP1-binding protein 16, Pc class 2 homolog, PC2, Polycomb 2 homolog | |
CBX4 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title