Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109482 Supplier Invitrogen™ Supplier No. PA5109482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVGLNLSHVR KRCLSETHGE REPCKKRLTA RSISTPTCLG GSPAAERPAD LPPAAALPQP EVILLDSDLD EPIDLRCVKT RSEAGEPPSS LQVKPETPAS AAVAVAAAAA PTTTAEKPPA EAQDEPAESL SEFKPFFGNI IITDVTANCL TVTFKEYVTV.

Polycomb group (PcG) proteins are chromatin-associated proteins which are important for embryonic and adult stem cell self-renewal and maintenance. At least two distinct human PcG complexes have been identified. Polycomb repressive complex I (PRC1) contains CBX4/Pc2, HPH1 and RING domain-containing proteins (RING1, RING2 and BMI1). CBX4 (Chromobox protein homolog 4 or PC2), which functions as a transcriptional suppressor, is a candidate of KyoT2-binding proteins. It is a E3 SUMO-protein ligase which facilitates SUMO1 conjugation. Depletion of CBX4 results in decreased cellular resistance to ionizing radiation. CBX4 is directly involved in the cellular response to DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CBX4
Gene Accession No. O00257, O55187
Gene Alias Cbx4; chromobox 4; chromobox homolog 4; chromobox homolog 4 (Pc class homolog, Drosophila); chromobox protein homolog 4; chromobox-like protein 4; E3 SUMO-protein ligase CBX4; E3 SUMO-protein transferase CBX4; hPc2; MPc2; NBP16; NS5ATP1-binding protein 16; Pc class 2 homolog; PC2; Polycomb 2 homolog
Gene Symbols CBX4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 401-560 of human CBX4 (NP_003646.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12418, 8535
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.