Learn More
Invitrogen™ CBX4 Polyclonal Antibody

Description
Immunogen sequence: SPKYNTWEPE ENILDPRLLI AFQNRERQEQ LMGYRKRGPK PKPLVVQVPT FARRSNVLTG LQDSSTDNRA KLDSGAEEK Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.
Specifications
Specifications
| Antigen | CBX4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CBX4 |
| Gene Accession No. | O00257 |
| Gene Alias | Cbx4; chromobox 4; chromobox homolog 4; chromobox homolog 4 (Pc class homolog, Drosophila); chromobox protein homolog 4; chromobox-like protein 4; E3 SUMO-protein ligase CBX4; E3 SUMO-protein transferase CBX4; hPc2; MPc2; NBP16; NS5ATP1-binding protein 16; Pc class 2 homolog; PC2; Polycomb 2 homolog |
| Gene Symbols | CBX4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.