Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS17376R

Description
Polycomb group (PcG) proteins are chromatin-associated proteins which are important for embryonic and adult stem cell self-renewal and maintenance. At least two distinct human PcG complexes have been identified. Polycomb repressive complex I (PRC1) contains CBX4/Pc2, HPH1 and RING domain-containing proteins (RING1, RING2 and BMI1). CBX4 (Chromobox protein homolog 4 or PC2), which functions as a transcriptional suppressor, is a candidate of KyoT2-binding proteins. It is a E3 SUMO-protein ligase which facilitates SUMO1 conjugation. Depletion of CBX4 results in decreased cellular resistance to ionizing radiation. CBX4 is directly involved in the cellular response to DNA damage.Specifications
CBX4 | |
Polyclonal | |
Unconjugated | |
CBX4 | |
Cbx4; chromobox 4; chromobox homolog 4; chromobox homolog 4 (Pc class homolog, Drosophila); Chromobox protein homolog 4; chromobox-like protein 4; E3 SUMO-protein ligase CBX4; E3 SUMO-protein transferase CBX4; hPc2; MPc2; NBP16; NS5ATP1-binding protein 16; Pc class 2 homolog; Pc2; Polycomb 2 homolog | |
Rabbit | |
Protein A | |
RUO | |
8535 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O00257 | |
CBX4 | |
KLH conjugated synthetic peptide derived from human CBX4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction