Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX5 Mouse anti-Human, Mouse, Clone:1B10B2, Abnova™
Mouse monoclonal antibody raised against recombinant human CBX5.
Supplier: Abnova Corporation MAB17166
Description
This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain (CSD) which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The encoded product is involved in the formation of functional kinetochore through interaction with essential kinetochore proteins. The gene has a pseudogene located on chromosome 3. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]Specifications
CBX5 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry The optimal working dilution should be determined by the end user. |
|
CBX5 | |
Mouse | |
100 μg | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1B10B2 | |
chromobox homolog 5 (HP1 alpha homolog, Drosophila) | |
In PBS (0.05% sodium azide) | |
HP1, HP1A | |
Recombinant protein corresponding to amino acid 1-191 of human CBX5 from E. coli. | |
23468 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction