Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX6 Antibody (8D10A3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | CBX6 |
---|---|
Clone | 8D10A3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
Description
CBX6 Monoclonal specifically detects CBX6 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
CBX6 | |
1 mg/mL | |
Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
chromobox homolog 6, chromobox protein homolog 6 | |
CBX6 | |
IgG1 | |
Protein A or G purified |
8D10A3 | |
Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
23466 | |
Purified recombinant fragment of human CBX6 (AA: 269-412) expressed in E. Coli. | |
Primary | |
43.9 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title