Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX7 Antibody (3H9D7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261724
Description
CBX7 Monoclonal specifically detects CBX7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CBX7 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
chromobox homolog 7, chromobox protein homolog 7 | |
Mouse | |
33.8 kDa | |
0.1 mL | |
23492 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
3H9D7 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
CBX7 | |
Purified recombinant fragment of human CBX7 (AA: 15-147) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction