Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBX7 Monoclonal Antibody (3H9C4)
GREENER_CHOICE

Catalog No. PIMA531668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531668 Supplier Invitrogen™ Supplier No. MA531668
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31668 has been tested in indirect ELISA.

The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3H9C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CBX7
Gene Accession No. O95931
Gene Alias 1600014J01Rik; AI851678; Cbx7; chromobox 7; chromobox homolog 7; chromobox protein homolog 7; chromobox-like protein 7; D15Ertd417e
Gene Symbols CBX7
Host Species Mouse
Immunogen Purified recombinant fragment of human CBX7 (AA: 15-147) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23492
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.