Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX7 Mouse anti-Human, Clone:3H9C4, Abnova™
Mouse monoclonal antibody raised against partial recombinant human CBX7.
Supplier: Abnova Corporation MAB17888
Specifications
CBX7 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
CBX7 | |
Recombinant protein corresponding to amino acids 15-147 of human CBX7. | |
23492 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
ELISA, Western Blot | |
3H9C4 | |
chromobox homolog 7 | |
In PBS (0.05% sodium azide) | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction