Learn More
CBX7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | CBX7 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2556
![]() |
Bethyl Laboratories, Inc
IHC00696T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2555
![]() |
Bethyl Laboratories, Inc
IHC00696 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Horse, Rabbit, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibb
Involved in maintaining the transcriptionally repressive state of genes. Modifies chromatin, rendering it heritably changed in its expressibility (By similarity). Regulator of cellular lifespan by maintaining the repression of CDKN2A, but not by inducing telomerase activity.Specifications
CBX7 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
chromobox protein homolog 7 | |
CBX7 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O95931 | |
23492 | |
Between 201 and 251 | |
Primary | |
4° C | |
CBX7 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.