Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX8 Monoclonal Antibody (1214CT171.154.107)
CBX8 Monoclonal Antibody (1214CT171.154.107)
Supplier: Thermo Scientific MA537659
Description
CBX8 Monoclonal Antibody for Western Blot, Flow

Specifications
CBX8 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9HC52 | |
CBX8 | |
KLH conjugated synthetic peptide between amino acids from the C-terminal region of human CBX8. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
1214CT171.154.107 | |
Unconjugated | |
CBX8 | |
CBX8; chromobox 8; chromobox homolog 8; chromobox homolog 8 (Pc class homolog, Drosophila); chromobox protein homolog 8; HPC3; mPc3; Pc class 3 homolog; Pc3; polycomb 3; polycomb 3 homolog; RC1; rectachrome 1 | |
Mouse | |
Protein G | |
RUO | |
57332 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction