Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX8 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI6G4, lyophilized, TrueMAB™
SDP

Catalog No. 501665142
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-5142 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-5142 Supplier OriGene Supplier No. CF800499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Chromobox homolog 8 (CBX8 or Pc3) is a member of Drosophila Polycomb group gene family. The polycomb group (PcG) genes are essential for maintece of appropriate expression patterns of developmental master regulators and thus are essential for proper development. Changes in expression of PcG proteins have been associated with cancer. CBX8 is involved in maintaining the transcriptionally repressive state of genes. It modifies chromatin, rendering it heritably changed in its expressibility. Structurally, CBX4 contains 1 chromo domain, which is 40 to 50 amino acids long.

Specifications

Antigen CBX8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6G4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CBX8
Gene Accession No. Q9HC52, Q9QXV1
Gene Alias PC3, RC1
Gene Symbols CBX8
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-260 of human CBX8 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303731, 30951, 57332, 608591
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.