Learn More
CBX8 Mouse anti-Human, Clone: OTI6D9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800491

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Chromobox homolog 8 (CBX8 or Pc3) is a member of Drosophila Polycomb group gene family. The polycomb group (PcG) genes are essential for maintece of appropriate expression patterns of developmental master regulators and thus are essential for proper development. Changes in expression of PcG proteins have been associated with cancer. CBX8 is involved in maintaining the transcriptionally repressive state of genes. It modifies chromatin, rendering it heritably changed in its expressibility. Structurally, CBX4 contains 1 chromo domain, which is 40 to 50 amino acids long.Specifications
CBX8 | |
Monoclonal | |
Unconjugated | |
CBX8 | |
PC3, RC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57332 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6D9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9HC52 | |
CBX8 | |
Human recombit protein fragment corresponding to amino acids 1-260 of human CBX8 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.